siRNA Details
| Gene Symbol: 4930447A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015396.1 | - | 1 | 150 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714949.1 | - | 146 | Chr. - - [-] on Build | ||

4930447A16Rik
Genomic Map