siRNA Details
| Gene Symbol: 4930445N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125720.1 | 3 | 853 | |||
| GenBank mRNA | AK015395.1 | - | 3 | 854 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930445N18Rik| Gene Symbol: 4930445N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125720.1 | 3 | 853 | |||
| GenBank mRNA | AK015395.1 | - | 3 | 854 | ||