siRNA Details
| Gene Symbol: 4930445K14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015392.1 | - | 1135 | |||
| AK142117.1 | - | 1112 | ||||
| BC037706.1 | - | 1109 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930445K14Rik| Gene Symbol: 4930445K14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015392.1 | - | 1135 | |||
| AK142117.1 | - | 1112 | ||||
| BC037706.1 | - | 1109 | ||||