siRNA Details
| Gene Symbol: Gm30790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038693.1 | - | 4 | 611 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB632473.1 | - | 4 | 611 | Chr. 16 - 76752113 - 76779092 [-] on Build GRCm38 | |

Gm30790
Genomic Map