siRNA Details
| Gene Symbol: 4933417E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040454.1 | 3 | 644 | |||
| GenBank mRNA | AK016843.1 | - | 3 | 645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933417E11Rik| Gene Symbol: 4933417E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040454.1 | 3 | 644 | |||
| GenBank mRNA | AK016843.1 | - | 3 | 645 | ||