siRNA Details
Gene Symbol: 4933417E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040454.1 | 3 | 1008 | |||
GenBank mRNA | AK016843.1 | - | 3 | 1009 | ||
AV280763.1 | - | 1 | 139 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933417E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040454.1 | 3 | 1008 | |||
GenBank mRNA | AK016843.1 | - | 3 | 1009 | ||
AV280763.1 | - | 1 | 139 |