siRNA Details
| Gene Symbol: 4933417D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045849.1 | 1 | 396 | |||
| GenBank mRNA | AK016841.1 | - | 1 | 397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933417D19Rik| Gene Symbol: 4933417D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045849.1 | 1 | 396 | |||
| GenBank mRNA | AK016841.1 | - | 1 | 397 | ||