siRNA Details
Gene Symbol: 1700097N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045287.1 | 4 | 694 | |||
GenBank mRNA | AK007099.1 | - | 2 | 193 | ||
AK018977.1 | - | 2 | 148 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700097N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045287.1 | 4 | 694 | |||
GenBank mRNA | AK007099.1 | - | 2 | 193 | ||
AK018977.1 | - | 2 | 148 |