siRNA Details
| Gene Symbol: 1700097N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045287.1 | 4 | 623 | |||
| GenBank mRNA | AK007099.1 | - | 2 | 122 | ||
| AK018977.1 | - | 2 | 77 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700097N02Rik| Gene Symbol: 1700097N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045287.1 | 4 | 623 | |||
| GenBank mRNA | AK007099.1 | - | 2 | 122 | ||
| AK018977.1 | - | 2 | 77 | |||