siRNA Details
| Gene Symbol: 1700121M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018973.1 | - | 1 | 323 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707326.1 | - | 1 | 323 | Chr. 15 - 99919874 - 99920299 [+] on Build GRCm38 | |

1700121M21Rik
Genomic Map