siRNA Details
| Gene Symbol: C230014O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048710.1 | - | 4 | 685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C230014O12Rik| Gene Symbol: C230014O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048710.1 | - | 4 | 685 | ||