siRNA Details
| Gene Symbol: 4633402D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014620.1 | - | 2 | 642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4633402D09Rik| Gene Symbol: 4633402D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014620.1 | - | 2 | 642 | ||