siRNA Details
| Gene Symbol: Deptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014624.1 | - | 1 | 1536 | ||
| AK171902.1 | - | 3702 | ||||
| AK172487.1 | - | 1 | 3112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Deptor| Gene Symbol: Deptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014624.1 | - | 1 | 1536 | ||
| AK171902.1 | - | 3702 | ||||
| AK172487.1 | - | 1 | 3112 | |||