siRNA Details
| Gene Symbol: Sh2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042476.1 | - | 819 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2d1a| Gene Symbol: Sh2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042476.1 | - | 819 | |||