siRNA Details
Gene Symbol: Gm28154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034407.1 | - | 1 | 726 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm28154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034407.1 | - | 1 | 726 |