siRNA Details
| Gene Symbol: Gm29729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039079.1 | - | 1 | 2672 | ||
| Gene Symbol: A230006K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046019.1 | - | 1 | 1934 | ||

Gm29729
Genomic Map