siRNA Details
| Gene Symbol: 9130015L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040499.1 | 3 | 252 | |||
| GenBank mRNA | AK018627.1 | - | 3 | 253 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720808.1 | - | 3 | 253 | Chr. 2 - 160016653 - 160040620 [+] on Build GRCm38 | |

9130015L21Rik
Genomic Map