siRNA Details
| Gene Symbol: 4930452A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045433.1 | 11 | ||||
| GenBank mRNA | AK016904.1 | - | 1 | 16 | ||
| AK019626.1 | - | 12 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714991.1 | - | 12 | Chr. 17 - 9750751 - 9775866 [-] on Build GRCm38 | ||

4930452A19Rik
Genomic Map