siRNA Details
| Gene Symbol: 4930452A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045433.1 | 3 | 401 | |||
| GenBank mRNA | AK019626.1 | - | 3 | 402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930452A19Rik| Gene Symbol: 4930452A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045433.1 | 3 | 401 | |||
| GenBank mRNA | AK019626.1 | - | 3 | 402 | ||