siRNA Details
| Gene Symbol: D130043K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019624.1 | - | 1 | 275 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714986.1 | - | 1 | 271 | Chr. 13 - 24845115 - 24845724 [+] on Build GRCm38 | |

D130043K22Rik
Genomic Map