siRNA Details
Gene Symbol: D130043K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019624.1 | - | 1 | 503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714986.1 | - | 1 | 499 | Chr. 13 - 24845115 - 24845724 [+] on Build GRCm38 |
