siRNA Details
| Gene Symbol: Gm10390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045793.1 | 3 | 3292 | |||
| GenBank mRNA | AK041808.1 | - | 3 | 3290 | ||
| AK169507.1 | - | 3 | 3298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10390| Gene Symbol: Gm10390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045793.1 | 3 | 3292 | |||
| GenBank mRNA | AK041808.1 | - | 3 | 3290 | ||
| AK169507.1 | - | 3 | 3298 | |||