siRNA Details
| Gene Symbol: 4933405E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045506.1 | 1 | 430 | |||
| GenBank mRNA | AK016664.1 | - | 1 | 433 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716353.1 | - | 1 | 433 | Chr. 11 - 54177938 - 54213290 [-] on Build GRCm38 | |

4933405E24Rik
Genomic Map