siRNA Details
| Gene Symbol: 4933405E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045506.1 | 3 | 883 | |||
| GenBank mRNA | AK016664.1 | - | 3 | 886 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933405E24Rik| Gene Symbol: 4933405E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045506.1 | 3 | 883 | |||
| GenBank mRNA | AK016664.1 | - | 3 | 886 | ||