siRNA Details
| Gene Symbol: Gm6297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077221.1 | 3 | 742 | |||
| NR_077226.1 | 3 | 904 | ||||
| GenBank mRNA | AK139286.1 | - | 1 | 1993 | ||
| AK139295.1 | - | 2 | 1429 | |||
| AK172127.1 | - | 1 | 1540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6297| Gene Symbol: Gm6297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077221.1 | 3 | 742 | |||
| NR_077226.1 | 3 | 904 | ||||
| GenBank mRNA | AK139286.1 | - | 1 | 1993 | ||
| AK139295.1 | - | 2 | 1429 | |||
| AK172127.1 | - | 1 | 1540 | |||