siRNA Details
| Gene Symbol: D530033B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021305.1 | - | 1 | 166 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730649.1 | - | 1 | 163 | Chr. 7 - 38182559 - 38182775 [-] on Build GRCm38 | |

D530033B14Rik
Genomic Map