siRNA Details
| Gene Symbol: 5430435K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017398.1 | - | 2 | 622 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717213.1 | - | 2 | 622 | Chr. 5 - 151252305 - 151259264 [+] on Build GRCm38 | |

5430435K18Rik
Genomic Map