siRNA Details
| Gene Symbol: 5430434N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017393.1 | - | 1 | 116 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717207.1 | - | 1 | 112 | Chr. 16 - 42510228 - 42510875 [-] on Build GRCm38 | |

5430434N17Rik
Genomic Map