siRNA Details
| Gene Symbol: 1700092K14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045930.1 | 2 | 133 | |||
| GenBank mRNA | AK007042.1 | - | 2 | 134 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707110.1 | - | 2 | 131 | Chr. 11 - 114198260 - 114199199 [+] on Build GRCm38 | |

1700092K14Rik
Genomic Map