siRNA Details
Gene Symbol: Bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032333.1 | - | 1 | 3187 | ||
AK079251.1 | - | 722 | ||||
AV341352.2 | - | 1 | 34 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032333.1 | - | 1 | 3187 | ||
AK079251.1 | - | 722 | ||||
AV341352.2 | - | 1 | 34 |