siRNA Details
| Gene Symbol: 1700071M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045444.1 | 1 | 339 | |||
| GenBank mRNA | AK006933.1 | - | 535 | |||
| AK133239.1 | - | 1 | 339 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706947.1 | - | 2 | 535 | Chr. 17 - 43591160 - 43592432 [-] on Build GRCm38 | |

1700071M16Rik
Genomic Map