siRNA Details
| Gene Symbol: 1700012J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005917.1 | - | 1 | 291 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700012J22Rik| Gene Symbol: 1700012J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005917.1 | - | 1 | 291 | ||