siRNA Details
| Gene Symbol: Clcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088307.1 | - | 426 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clcn3| Gene Symbol: Clcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088307.1 | - | 426 | |||