siRNA Details
| Gene Symbol: Kank4os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040437.1 | 3 | 833 | |||
| GenBank mRNA | AK043321.1 | - | 3 | 834 | ||
| AK166708.1 | - | 3 | 804 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kank4os| Gene Symbol: Kank4os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040437.1 | 3 | 833 | |||
| GenBank mRNA | AK043321.1 | - | 3 | 834 | ||
| AK166708.1 | - | 3 | 804 | |||