siRNA Details
| Gene Symbol: Gm38397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089337.1 | - | 4 | 454 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY749581.1 | - | 4 | 451 | Chr. 13 - 110903501 - 110906580 [+] on Build GRCm38 | |

Gm38397
Genomic Map