siRNA Details
| Gene Symbol: 6720482G16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020168.1 | - | 1 | 382 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719454.1 | - | 1 | 382 | Chr. 3 - 58526930 - 58527602 [+] on Build GRCm38 | |

6720482G16Rik
Genomic Map