siRNA Details
| Gene Symbol: Gm31135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082314.1 | - | 605 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB650060.1 | - | 6 | 605 | Chr. 8 - 8045050 - 8484721 [-] on Build GRCm38 | |

Gm31135
Genomic Map