siRNA Details
| Gene Symbol: Gm31641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087149.1 | - | 626 | |||
| BC099513.1 | - | 5 | 546 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK791783.1 | - | 5 | 567 | Chr. 16 - 95185235 - 95259627 [-] on Build GRCm38 | |

Gm31641
Genomic Map