siRNA Details
| Gene Symbol: 1810035K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019027.1 | - | 2 | 163 | ||
| AV056323.1 | - | 1 | 108 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707721.1 | - | 2 | 163 | Chr. 19 - 58695521 - 58696181 [+] on Build GRCm38 | |

1810035K13Rik
Genomic Map