siRNA Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF498302.1 | - | 3 | 243 | ||
| EU434919.1 | - | 9 | 817 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rian| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF498302.1 | - | 3 | 243 | ||
| EU434919.1 | - | 9 | 817 | |||