siRNA Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF498302.1 | - | 2 | 166 | ||
| EU434919.1 | - | 8 | 740 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG694943.1 | - | 2 | 410 | Chr. 12 - 109633959 - 109643464 [+] on Build GRCm38 | |

Rian
Genomic Map