siRNA Details
Gene Symbol: Rian | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF498302.1 | - | 2, 3 | 218 | ||
EU434919.1 | - | 8, 9 | 792 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG694943.1 | - | 2, 3 | 462 | Chr. 12 - 109633959 - 109643464 [+] on Build GRCm38 |
