siRNA Details
| Gene Symbol: 2900083M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019360.1 | - | 1 | 130 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712779.1 | - | 1 | 128 | Chr. 2 - 56265846 - 56266061 [+] on Build GRCm38 | |

2900083M14Rik
Genomic Map