siRNA Details
| Gene Symbol: Frmpd1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040666.1 | 1 | 13 | |||
| GenBank mRNA | AK019369.1 | - | 1 | 15 | ||
| BY712818.1 | - | 11 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmpd1os| Gene Symbol: Frmpd1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040666.1 | 1 | 13 | |||
| GenBank mRNA | AK019369.1 | - | 1 | 15 | ||
| BY712818.1 | - | 11 | ||||