siRNA Details
Gene Symbol: Frmpd1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040666.1 | 3 | 329 | |||
GenBank mRNA | AK019369.1 | - | 3 | 331 | ||
BY712818.1 | - | 3 | 327 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Frmpd1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040666.1 | 3 | 329 | |||
GenBank mRNA | AK019369.1 | - | 3 | 331 | ||
BY712818.1 | - | 3 | 327 |