siRNA Details
| Gene Symbol: Gm38509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110488.1 | 1 | 12 | |||
| GenBank mRNA | AW988896.1 | - | 1 | 12 | ||
| BC051220.1 | - | 1 | 21 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm38509| Gene Symbol: Gm38509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110488.1 | 1 | 12 | |||
| GenBank mRNA | AW988896.1 | - | 1 | 12 | ||
| BC051220.1 | - | 1 | 21 | |||