siRNA Details
| Gene Symbol: Macrod2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031201.1 | - | 8 | 2863 | ||
| Gene Symbol: Macrod2os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158168.1 | - | 2 | 2540 | ||

Macrod2os1
Genomic Map