siRNA Details
| Gene Symbol: Gm19689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045094.1 | 1 | 70 | |||
| GenBank mRNA | AK051223.1 | - | 1 | 71 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB655718.1 | - | 1 | 71 | Chr. 17 - 83036961 - 83078225 [-] on Build GRCm38 | |

Gm19689
Genomic Map