siRNA Details
Gene Symbol: Gm19689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045094.1 | 4 | 1032 | |||
GenBank mRNA | AK051223.1 | - | 4 | 1033 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm19689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045094.1 | 4 | 1032 | |||
GenBank mRNA | AK051223.1 | - | 4 | 1033 |