siRNA Details
| Gene Symbol: Gm19689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045094.1 | 4 | 1131 | |||
| GenBank mRNA | AK051223.1 | - | 4 | 1132 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19689| Gene Symbol: Gm19689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045094.1 | 4 | 1131 | |||
| GenBank mRNA | AK051223.1 | - | 4 | 1132 | ||